VWR® Bottle Tote™ Safety Carrier
Catalog # 470204-578
Supplier: VWR International
Specifications
- Description:VWR® Bottle Tote, Safety Carrier
- Environmentally Preferable:
- Cat. No.:470204-578
- Supplier no.:B-102
Specifications
About this item
Designed for safe transport of acids, alkalies, and solvents, a centering ring in the base assures 1" clearance between bottle and wall of the carrier.
- Can be steam sterilized
- Centering ring assures proper clearance for safe transport
Molded-in handle provides comfortable grip and will withstand 80lb. pull. Black carrier can be steam sterilized and accommodates 2.5 L (5-pint) or 4L (1-gallon) bottles.