Fertile Chicken Eggs
Supplier: Avantor
Certificates
About this item
Drive home the concepts of embryology by hatching these viable chicken eggs in your classroom incubator.
- Fertile broiler eggs are perfect for incubation and hatching at home or school
- Will fit in most incubators
- Hatch in approximately 21 days when kept at 99 °F
- Produce broiler chicks with gray and red variations
Three weeks notice are required to meet your requested delivery date. Eggs are specially packaged to prevent breakage and are guaranteed fresh and fertile. Shipping and incubation conditions will affect the success of your hatch so we cannot guarantee complete hatch.
Frequently Bought Together