Vernier® LabQuest Mini® Data Logger
Vernier LabQuest Mini
Catalog # 470017-584
Supplier: Vernier Software & Technology
Supplier #: LQ-MINI
Jump to:
Product Details & Documents
LabQuest Mini is powerful, affordable, and easy to use. It brings the power of Vernier's award-winning LabQuest technology to teachers who don't need the versatility of a standalone device. It is the perfect solution for educators collecting data with a windows or macintosh computer.
- Maximum sampling rate: 100 kHz
- Five sensor ports
- Over 70 compatible sensors
- Analog inputs: 3
- Digital inputs: 2
- Logger Pro 3, logger lite, and graphical analysis for chrome compatible
LabQuest Mini software has unparalleled power, analysis, and curricular support. It is for educational use only. USB used for computer connection, full speed of 2.0.
Maximum sampling rate is 100000 samples per second. Power is supplied by the USB port of the computer - auxiliary AC power or batteries are not required.
Delivery information: It includes computer connection (USB) cable and user manual.
Specifications
- Description:Vernier LabQuest Mini