ContactUsLinkComponent
Vernier® Color Mixer Kit
Vernier® Color Mixer Kit
Catalog # 470017-518
undefined
Vernier® Color Mixer Kit
Catalog # 470017-518
Supplier Number:  CM-OEK
Specifications
  • Description:
    Color Mixer Kit
  • Environmentally Preferable:
  • Cat. No.:
    470017-518
  • Supplier no.:
    CM-OEK
Specifications
About this item
Students discover wavelengths of visual light as they create the colors of the rainbow or even customize their own unique tint.
  • Adjustable LED Lights
  • Additive or Subtractive Hue Mixing
  • Power Supply Included
The intensity of the red, green, and blue LED lights can be adjusted from the light source for simple additive or subtractive hue mixing and fun experimentation in color.
This kit is for educational use only.
Ordering information: The kit consists of an LED illuminator with primary colors, a lens, a double-sided screen, and a power supply. It requires the Vernier Dynamics Track, which is sold separately.