ContactUsLinkComponent
Vernier® UV Sensors
Vernier® UV Sensors
Certificates
undefined
Vernier® UV Sensors
Specifications
  • Catalog No:
  • 470017-564
  • 470017-562
  • Resolution:
  • 0.25 mW/m²
  • 5.0 mW/m²
  • Response Time:
  • Approximately 2 s to 95% of final reading
  • Approximately 2 s to 95% of final reading
  • Dimensions:
  • Ø 20×210 mm
  • Ø 20×210 mm