ContactUsLinkComponent
Using Chemical Reactions to Reduce Waste Kit
Using Chemical Reactions to Reduce Waste Kit
Catalog # 470113-078
Supplier:  Lab-Aids
undefined
Using Chemical Reactions to Reduce Waste Kit
Catalog # 470113-078
Supplier:  Lab-Aids
Supplier Number:  441S
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Using Chemical Reactions to Reduce Waste Kit
  • No. of lab groups:
    6
  • Cat. No.:
    470113-078
  • Includes:
    Teacher's Guide, Student Guides and Worksheets, SDS
  • Supplier no.:
    441S
Specifications
About this item
Teach Students The Importance of Efficient Waste Disposal
  • Students reclaim waste that could harm the environment
  • Students make decisions about complex issues involving trade-offs
  • Requires one to two 50 minute class periods
Wastes from industrial processes sometimes contain toxic or valuable substances that can be reclaimed and then recycled or disposed of safely. Students will work in groups to reclaim copper from a liquid "waste" which contains dissolved copper chloride as well as measure the effectiveness of copper removal from three different metal replacement reactions. Students will then compare the cost, effectiveness and safety information for each replacement reaction to decide which metal is most efficient in the copper removal of the waste.
Ordering information: Materials Include Chemplates®, Aluminum washers, Iron washers, Zinc washers, Waste copper chloride solution, 15-mL drop control bottles, Plastic spoons, Dropper, Transparency: Wastewater Discharge Limits.