Universal Diamond ATR
470203-808
- Description:Universal Diamond ATR for Spectrum Two™
- Cat. No.:470203-808
- Supplier no.:L1600107
Ensures Highly Reproducible Results
This universal ATR is an excellent general accessory for FT-IR analysis of solids and liquids using any spectrometer. Setup is simple with plug-and-play operation that requires no alignment and live force readout as samples are positioned. Produce high quality spectra through the use of a pressure arm allowing good contact of the sample with the diamond crystal. The pressure arm force indicator ensures first-class sample-to-sample and operator-to-operator reproducibility. A secure vapor-sealed lid ensures samples are protected from contamination, for accurate results with every use. Not for sale in Canada.