ContactUsLinkComponent
Ungraduated Thermometers
  470104-552
 :  Echo
undefined
Ungraduated Thermometers
  470104-552
 :  Echo
 :  BR/2496 PU
  • Description:
    Ungraduated Thermometers
  • Cat. No.:
    470104-552
  • Supplier no.:
    BR/2496 PU
Make your own thermometers.
Precision made 12.5" glass tubes with a range of 0 to 245 °F. Filled with a special non-toxic oil that will not fade or separate. Pre-tested for accuracy. Package of six.