Order Entry
United States
Orders LinkContactUsLinkComponent
Two-Point Aesthesiometer

Two-Point Aesthesiometer

Two-Point Aesthesiometer, 15 cm, 2 cm
Catalog # 470019-282
Supplier #:  12-1480
Two-Point Aesthesiometer
Catalog # 470019-282
Two-Point Aesthesiometer
Two-Point Aesthesiometer

Two-Point Aesthesiometer, 15 cm, 2 cm

Price
$59.40
Log in for availability
Each Case of 10
Product Details & Documents

Evaluate Cutaneous Sensitivity And Touch Threshold

The sliding scale measures the ability of a subject to distinguish between two points of stimuli on the skin, while the plastic points minimize the influence of temperature. The scale, printed directly on the device, measures up to 14 cm in 1 mm increments.

Specifications
  • Description:
    Two-Point Aesthesiometer
  • Length:
    15 cm
  • Width:
    2 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.