ContactUsLinkComponent
Tryptic Soy Broth
Tryptic Soy Broth
Catalog # 470015-844
Supplier:  Avantor
CAS Number:  
Tryptic Soy Broth
Catalog # 470015-844
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    Dehydrated Tryptic Soy Broth
  • Cat. No.:
    470015-844
  • Supplier no.:
    WARD470015-844
Specifications
About this item
Soybean-casein digest medium. Supports the growth of a variety of microorganisms.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.