Trypanosoma rhodesiense Slide
Catalog # 470181-914
Supplier: Avantor
Specifications
- Description:Trypanosoma rhodesiense Slide
- Environmentally Preferable:
- Cat. No.:470181-914
- Stain:Giemsa
- Supplier no.:WARD470181-914
- Orientation:Smear
Specifications
About this item
Human parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
From Mammal blood. Causes Rhodesian sleeping sickness.