Order Entry
United States
Orders LinkContactUsLinkComponent
Vernier® Tris-Compatible Flat pH Sensor

Vernier® Tris-Compatible Flat pH Sensor

Vernier tris-compatable flat pH sensor (BTA), 12 mm, BNC, 1 m coaxial cable, PC, 5...80 °C, 0 - 14
Catalog # 470017-604
Supplier #:  FPH-BTA
Vernier® Tris-Compatible Flat pH Sensor
Catalog # 470017-604
Vernier® Tris-Compatible Flat pH Sensor
Vernier® Tris-Compatible Flat pH Sensor

Vernier tris-compatable flat pH sensor (BTA), 12 mm, BNC, 1 m coaxial cable, PC, 5...80 °C, 0 - 14

Price
$179.99
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The Tris-Compatible Flat pH Sensor measures the pH of a solution or semisolid.

  • Durable and easier to maintain than other pH sensors
  • Easy to clean

Unlike other pH sensors, this one has a flat glass membrane versus the typical glass bulb, making this sensor more sturdier and easier to clean while allowing for smaller sample sizes or flat surface measurements. This sensor is compatible with Tris buffers and any solution containing sulfides or proteins as it features a sealed, gel-filled, double-junction electrode Use this in conjunction with the Vernier LabQuest, LabQuest Mini, Vernier LabPro, Vernier Go! Link, Vernier EasyLink, Vernier SensorDAQ, and CBL 2.

Electrode type: Double-junction, sealed, gel-filled, Ag/AgCl reference, polycarbonate body
Membrane style: Flat glass
Storage solution: pH 4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution)
Cable: 1 meter coaxial cable with BNC connector
Impedance: 20 kΩ at 25ºC
Response time: 98% of full response in 30 s at 25 ºC

For educational use only.

Specifications
  • Ø:
    12 mm
  • Temperature range:
    5...80 °C
  • Description:
    Vernier tris-compatable flat pH sensor (BTA)
  • pH range:
    0 - 14
  • Body material:
    PC
  • Connector:
    BNC, 1 m coaxial cable
  • Cat. No.:
    470017-604
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.