Orders LinkContactUsLinkComponent
Treadprint™ Kit
Treadprint™ Kit
Treadprint™ Kit, 4
Catalog # 470000-072
Supplier:   FORENSICS SOURCE
Supplier #:  1008056
Treadprint™ Kit
Catalog # 470000-072
Supplier:  FORENSICS SOURCE
Treadprint™ Kit
Treadprint™ Kit
Treadprint™ Kit, 4
Price
$162.89
Log in for availability
Kit of 1
Product Details & Documents
A Convenient Method for Recording Tire Tread Standards
  • Make Impressions Like Forensic Experts
Tire tracks can be crucial evidence in a criminal case, so the impression and interpretation of treads is an important technique to master. This kit quickly and easily records impressions without requiring further development. Apply ink to the surface of a mounted tire with the included applicator, then roll the tire over the pre-cut panels for resulting tracks in black ink that dry immediately. Sixteen pre-cut panels are provided, enough to record the full rotation of 4 tires, size 205/R15. Panel size: 10" x 24".
Note: This product is available to US customers only
Specifications
  • Description:
    Treadprint™ Kit
  • No. of lab groups:
    4