Orders LinkContactUsLinkComponent
Trajectory Kit
Trajectory Kit
Supplier:   FORENSICS SOURCE
Trajectory Kit
Supplier:  FORENSICS SOURCE
Certificates
Trajectory Kit
3 Options Available Below
Product Details & Documents
Includes Everything Needed to Measure Trajectories at Crime Scenes.
  • Reuseable Materials
  • Math and Physics Intensely Used
Accurately measure angles and slopes every time with professional forensics equipment, including four 18L×¹/₄" diameter, yellow aluminum protrusion rods with connectors, to provide sufficient length for easy trajectory measurements; two 1" long and two 2" long spacer cones for placement and centering of protrusion rods in various sized holes; and a level and angle finder that is easily mounted to protrusion rods or the included aluminum tripod mount with universal thread. Available to U.S. customers only, similar product will ship elsewhere.
Product Family Options
Product Information
  • No. of lab groups
    Availability
    Price
  • 470016-328
    Trajectory kit
    Trajectory kit
    1
    Log in for availability
    Log in for availability
    Each
    Each - $354.55
    $354.55
    Each
       $354.55
    Supplier Number: 1008064
  • 470334-330
    Protrusion rods, small (0.22 cal)
    Protrusion rods, small (0.22 cal)
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $86.75
    $86.75
    Pack of 4
       $86.75
    Supplier Number: 1007801
  • 470334-328
    Protrusion rods (0.32 cal+)
    Protrusion rods (0.32 cal+)
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $105.60
    $105.60
    Pack of 4
       $105.60
    Supplier Number: 1007800