High Speed Rotary Tool Kit
470100-966
: Avantor
- Description:High Speed Rotary Tool Kit
- Environmentally Preferable:
- Cat. No.:470100-966
- Supplier no.:4000-2/30
This kit features a digital rotary tool that delivers variable speeds of 5,000 to 35,000 rpm, along with 30 assorted bits and a circle cutter/straight edge guide. Accessories include a durable storage case, instruction manual and a keyless chuck.