Tissues and Organs Slide Set
470183-576
: Avantor
- Type:Cellular Development
- Description:Tissues and Organs Slide Set
- No. of slides:10
- Cat. No.:470183-576
- Supplier no.:WARD470183-576
Illustrate Differences Between Tissues and Organ Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
All slides are prepared as sections unless otherwise specified.
: Set includes Tissues: Epithelium composite; Cartilage composite; Mouse tail, variety of tissues: bone, muscle, tendon, nerve (cs); Muscle types, composite; Organs: Brain composite; Salivary glands, composite; Esophagus, composite (cs); Stomach, composite; Small intestine, composite; and Ovary, mature (cs).