ContactUsLinkComponent
Large Tinted Desiccator Cabinet
  470121-956
 :  Bel-Art
Large Tinted Desiccator Cabinet
  470121-956
 :  Bel-Art
 :  H42053-0001_SE
  • Description:
    Tinted Medium Desiccator Cabinet
  • Cat. No.:
    470121-956
  • Supplier no.:
    H42053-0001
Made of smoke tinted glass. Door has top single roll latch. Four sets of shelf rails allow for low or high objects. Includes small tray for silica gel and two vented shelves. InsideDimensions: 9" L x 8" D x 9" H (23 x 23 x 20 cm).Overall dimensions: 10" L x 10" D x 10" H (25 cm).