ContactUsLinkComponent
TI-89 Titanium Graphing Calculator
TI-89 Titanium Graphing Calculator
Catalog # 470151-722
Supplier:  D&H
undefined
TI-89 Titanium Graphing Calculator
Catalog # 470151-722
Supplier:  D&H
Supplier Number:  TI89TITANIUM
Specifications
  • Description:
    TI-89 Titanium graphing calculator
  • Cat. No.:
    470151-722
  • Supplier no.:
    89T/TBL/1L1
Specifications
About this item
The TI-89 Titanium features 16 preloaded apps, 188KB of RAM, 2.7MB of flash ROM, a built-in USB port with cable and an operating manual on CD.
  • Includes connectivity cable and Connect™ software for information sharing
  • Requires 4 "AAA" batteries (not included) and features automatic power down
Other features include: up to 99 graphing equations defined and saved for each graphing mode, numeric evaluation of functions in tables and data variable format, and interactive analysis of function values, roots, maximums, minimums, integrals, derivatives, intersections, inflection points and arc lengths.