ContactUsLinkComponent
TI-36X Pro Calculator
TI-36X Pro Calculator
Catalog # 470349-978
Supplier:  D&H
undefined
TI-36X Pro Calculator
Catalog # 470349-978
Supplier:  D&H
Supplier Number:  TI36XPRO
Specifications
  • Description:
    TI-36X pro scientific calculator
  • Environmentally Preferable:
  • Cat. No.:
    470349-978
Specifications
About this item
Engineering/Scientific Calculator
  • One- and two- variable statistics
  • Display a defined function in a tabular form
  • Perform vectors and matrices using a vector and matrix entry window
Ideal for many types of math, science, and engineering classes. The Multiview™ four line display shows multiple calculations at the same time on screen. Choose from three solvers: numeric equation, polynomial, and system of linear equations. Determine the numeric derivative and integral for real functions. Select degrees/radians, floating/fix, number format modes.