Orders LinkContactUsLinkComponent
This Dynamic Planet Map
This Dynamic Planet Map
This Dynamic Planet Map
Catalog # 470023-416
Supplier #:  206335
This Dynamic Planet Map
Catalog # 470023-416
Supplier:  
This Dynamic Planet Map
This Dynamic Planet Map
This Dynamic Planet Map
Discontinued
Product Details & Documents
Detail Earth’s Dynamic Processes Through Geologic Time
Display Earth’s major lithospheric plates, the location of over 1,500 volcanoes active during the last 10,000 years, over 24,000 earthquake epicenters from events largely recorded between 1960 and 1990, and the sites of approximately 140 major impact craters scattered across the Earth’s surface. Clear and concise text and diagrams at the base of the map further explain geologic events of Earth’s history. Map scale: 1:30,000,000. Size: 41" x 58".
Specifications
  • Description:
    This Dynamic Planet Map