ContactUsLinkComponent
Thioglycollate Medium
Thioglycollate Medium
Catalog # 470180-662
Supplier:  Avantor
CAS Number:  
undefined
Thioglycollate Medium
Catalog # 470180-662
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    Prepared Thioglycollate Medium Tubes
  • Cat. No.:
    470180-662
  • Supplier no.:
    WARD470180-662
Specifications
About this item
Without indicator. For cultivation of anaerobic species and Clostridium. Suitable for fermentation studies where no indicator is required.