The Sheep Brain, A Basic Guide
Catalog # 470022-686
Supplier: A.J. Kirby
Specifications
- Description:The Sheep Brain, A Basic Guide
- Length:20 cm
- Cat. No.:470022-686
- No. of pages:24
- Width:27 cm
- Supplier no.:321139
Specifications
About this item
C.H. Vanderwolf and Richard K. Cooley
Sheep brains have become a good choice to study neuroanatomy in a wide variety of classrooms because they are large enough to be easily dissected and readily available at a reasonable cost. These study guides provide a series of photographs taken from several different views. The photos are accurate and easy to read. The Photographic Series also contains pictures of other common lab animals, as well as photos and diagrams describing the central nervous system in the appendixes. Spiral bound.