Termite, Worker Slide
Termite, Worker, Whole mount, Unstained
470181-612
: Avantor
Illustrates Body Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Worker termite demonstrating general body structures and biting mouthparts. Highly cooperative social structure.
- Description:Termite, Worker
- Stain:Unstained
- Orientation:Whole mount