ContactUsLinkComponent
Ward's® Live Termite Flagellates (Trychonympha and Pyrsonympha)
Ward's® Live Termite Flagellates (Trychonympha and Pyrsonympha)
Catalog # 470180-128
Supplier:  Avantor
undefined
Ward's® Live Termite Flagellates (Trychonympha and Pyrsonympha)
Catalog # 470180-128
Supplier:  Avantor
Specifications
  • Description:
    Termite Flagellates (Small)
  • No. of students:
    30
  • Cat. No.:
    470180-128
  • Packaging:
    Jar
  • Media:
    Live termites
  • Supplier no.:
    WARD470180-128
Specifications
About this item
Termite flagellates live in the intestine of Zootermopsis termites as symbiotic organisms. They can be easily isolated and observed.
  • Demonstrate symbiosis in your classroom
  • No reproductive termites are sent (we do not send queens)
Flagellate protozoa including the genera Trichonympha and Pyrsonympha live in the intestinal tract of dampwood termites (Zootermopsis sp.) There they live, protected from the elements, and provided with food; in return they help the termite break down the cellulose in the wood they eat.
Show your students this relationship by allowing them to observe the termites' feeding behaviors, and then isolate the symbionts.
No reproductive termites are sent with the culture. We do not send queens.
Ordering information: These products contain living or perishable materials.
No reproductive termites are sent with the culture. We do not send queens.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Ward's holds valid USDA permits allowing us to ship to most continental US states. Please call Customer Service for permit details.