Specifications
- Type:Human Body
- Description:Teeth & Gums Microslide
- Cat. No.:470105-400
- Supplier no.:T-220
Specifications
About this item
Explore oral hygiene in a new way
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
In this lesson students explore teeth and gums, the reasons for and importance of good dental hygiene, and what can go wrong if students do not practice good dental habits. Further, students will see how the function of teeth can be deduced from their shape.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Incisors
Canines - cs (1.3X)
Molars
Molar - cs (1.3X)
Cavity
Periodontal disease
Teeth with plaque deposits
Dental flossing