Tapeworm, Comparative Proglottids Slide
Catalog # 470182-072
Supplier: Avantor
Specifications
- Description:Tapeworm, Comparative Proglottids
- Cat. No.:470182-072
- Stain:Schneider's
- Supplier no.:WARD470182-072
- Orientation:Whole mount
Specifications
About this item
Proglottid Comparison
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mature proglottids of Taenia and Dipylidium are mounted on one slide.