ContactUsLinkComponent
Taenia pisiformis, Mature Proglottid Slide
Taenia pisiformis, Mature Proglottid Slide
Catalog # 470176-918
Supplier:  Avantor
undefined
Taenia pisiformis, Mature Proglottid Slide
Catalog # 470176-918
Supplier:  Avantor
Specifications
  • Description:
    Taenia pisiformis, Mature Proglottid
  • Environmentally Preferable:
  • Cat. No.:
    470176-918
  • Stain:
    Schneider's and Fast Green
  • Supplier no.:
    WARD470176-918
  • Orientation:
    Whole mount
Specifications
About this item
Endoparasitic Tapeworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mature proglottid of Taenia pisiformis, showing uterus, ovary, testes, sperm duct, and genital pore.