Orders LinkContactUsLinkComponent
Swivel Type Support Clamp
Swivel Type Support Clamp
Swivel Type Support Clamp
Catalog # 470123-516
Supplier:   Wards
Supplier #:  S78447-50
Swivel Type Support Clamp
Catalog # 470123-516
Supplier:  Wards
Swivel Type Support Clamp
Swivel Type Support Clamp
Swivel Type Support Clamp
Clearance
$34.80
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
A compact aluminum clamp for holding a 9.5 mm rod at any angle with a 12.7 mm rod.
Particularly useful for positioning equipment in the physics laboratory.
Specifications
  • Description:
    Swivel Type Support Clamp