Sun, Seasons, and Constellations Model Set
Product Details & Documents
Demonstrate How Earth’s Motions Affect Seasons, Climate, and Visible Constellations.
- Raised-Relief Globe Included
- Instructions, Lamp, and LCD Thermometers Included
- Students Experiment with Earth's Temperature Variations and More
Using a raised-relief globe, an infrared lamp, and LCD thermometers, your students can experiment with Earth’s seasonal and regional temperature variations, equinox and solstice, and rotation and orbit. The model also includes constellation drawings to illustrate how the constellations appear to change relative to Earth’s motion.
The kit includes a 12" raised-relief globe, one infrared lamp and reflector, three LCD thermometers, and instructions. A support stand for the lamp, required, is available separately.
Documents
WARD470004-620 2016-09-02.pdf
Additional Product Details
Specifications
- Description:Sun, Seasons, and Constellations Model Set
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- Triangular Support Stand with 8 Legs