[NA_SciEd] CENCO® String Vibrator
110V
Catalog # 470110-622
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WAVE GENERATOR PEARSONTIFICS Catalog #470212-494
Product 470212-494 is similar to 470110-622 but manufacturer and specifications may vary.
Product Details & Documents
Specifications
- Input voltage:110V
Accessories
Product Information
- Catalog #IncludesAvailabilityPrice
- Mass Set of 8, LabwareMass Set of 8, Labware470017-100(1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 gLog in for availabilityEach - $35.50
Product Information
- Catalog #WeightAvailabilityPrice
- Mass Hanger, 500 g Capacity
Product Information
- Catalog #ØAvailabilityPrice
- Clamp Pulley
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



