Orders LinkContactUsLinkComponent
Prepared Starch Agar Media
Prepared Starch Agar Media
Starch Agar, Plate
Catalog # 470180-696
Supplier #:  CM466-P25
Prepared Starch Agar Media
Catalog # 470180-696
CAS Number:  
Prepared Starch Agar Media
Prepared Starch Agar Media
Starch Agar, Plate
CAS Number:  
Price
$35.95
Log in for availability
Pack of 10
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
This media can be used to determine if a microorganism is capable of hydrolyzing starch as a source of energy for growth.
  • Sterile, pre-prepared media; just inoculate and incubate.
  • When used with an iodine solution, works as a test for alpha-amylase.
After the organism is plated onto the starch agar plate, an organism tests positive for alpha-amylase when, in the presence of an iodine solution, a blue-black color forms in the medium.
Each bottle pours approximately 10 Petri dishes. Simply melt in a microwave oven, water bath or autoclave. Once cool enough to handle, pour into sterile Petri dish plates
Documents
Specifications
Specifications
  • Pack type:
    Plate
  • Description:
    Starch Agar