Stainless Steel Cylinder Weight Set
Catalog # 470200-628
Supplier: Troemner
Specifications
- Description:Stainless Steel Cylinder Weights, Set of 13, 1 g to 1000 g
- Cat. No.:470200-628
- Supplier no.:301S
Specifications
About this item
Weight sets are fitted in a sturdy styrene storage block.
- Includes Storage Box
- Stainless Steel Construction
- Choice of Two Sets
The set of 12 includes one each of 1 g, 5 g, 20 g, 50 g, 200 g, and 500 g weights plus two each of 2 g, 10 g, 100 g weights.
The set of 13 includes all the same weights as the set of 12 plus one 1000 g weight.