Order Entry
United States
ContactUsLinkComponent
Square Battery Jar
Square Battery Jar
Catalog # 470103-992
Supplier:  Avantor
undefined
Square Battery Jar
Catalog # 470103-992
Supplier:  Avantor

Specifications

  • Description:
    Square Battery Jar
  • Cat. No.:
    470103-992
  • Supplier no.:
    619-4

Specifications

About this item

Square, flint glass battery jar with ground rim.

Capacity: 1.7 L
Size: 10 cm dia. x 20 cm H