ContactUsLinkComponent
Spinal Cord (Human), Thoracic Level Slide
Spinal Cord (Human), Thoracic Level Slide
Catalog # 470176-968
Supplier:  Avantor
Spinal Cord (Human), Thoracic Level Slide
Catalog # 470176-968
Supplier:  Avantor
Specifications
  • Description:
    Spinal Cord (Human), Thoracic Level
  • Cat. No.:
    470176-968
  • Stain:
    Hematoxylin and Eosin
  • Supplier no.:
    WARD470176-968
  • Orientation:
    Cross section
Specifications
About this item
Central Nervous System
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Thoracic level cross section showing white and grey matter.