Replacement Accesssories for Spectra of the Elements Kit
Catalog # 470111-390
Supplier: The Science Source
Specifications
- Description:Spectra of the Elements Kit, Replacement Hand Spectroscopes w/Gratings
- Cat. No.:470111-390
- Supplier no.:15615
Specifications
About this item
The replacement chemicals kit contains 5g each of calcium chloride, cupric chloride, cupric nitrate, lithium chloride, potassium chloride, potassium nitrate, sodium bicarbonate, sodium chloride, strontium chloride and strontium nitrate.
An alcohol or bunsen burner is required.