Sound Tubes
Catalog # 470012-634
Supplier: Avantor
Specifications
- Description:Sound Tubes
- Cat. No.:470012-634
- Supplier no.:WARD470012-634
Specifications
About this item
Relate Motion to Sound
- Includes 10 assorted color tubes
- 29 1/2" long
Feel the effect of sound and spinning speed with these ten assorted plastic tubes. The faster the tube is spun, the more changes can be observed in the sound. If the tubes are cut to different lengths, the sounds and effects will also vary