Order Entry
United States
ContactUsLinkComponent
Somso® Torso with Head
Somso® Torso with Head
Catalog # 470007-332
Supplier:  MARCUS SOMMER
undefined
Somso® Torso with Head
Catalog # 470007-332
Supplier:  MARCUS SOMMER
Supplier Number:  AS 21

Specifications

  • Description:
    Torso and head
  • Height:
    89 cm
  • Length:
    26 cm
  • Cat. No.:
    470007-332
  • Width:
    39 cm
  • Supplier no.:
    AS 21

Specifications

About this item

Somso® premium quality models.

  • Natural size
  • Five year warranty

Separates into 15 parts: left half of brain, eye with muscles and optic nerve, halves of the lung (2), heart (2), liver, stomach, small and large intestine (3), opening appendix, half of right kidney, half of bladder, torso.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.