Order Entry
United States
ContactUsLinkComponent
Somso® Fertilization of the Angiosperms
Somso® Fertilization of the Angiosperms
Catalog # 470029-844
Supplier:  MARCUS SOMMER
undefined
Somso® Fertilization of the Angiosperms
Catalog # 470029-844
Supplier:  MARCUS SOMMER
Supplier Number:  BOS 19

Specifications

  • Description:
    Angiosperm fertilization
  • Height:
    66 cm
  • Length:
    14 cm
  • Cat. No.:
    470029-844
  • Width:
    30 cm
  • Supplier no.:
    BOS 19

Specifications

About this item

Somso® premium quality models.

  • Five year warranty

Polygonum-type, enlarged 300 times, in SOMSO-Plast®. After Prof. Dr. W. Jung. Longitudinal section showing ovary with germinating pollen grains and embryo sac, stamen with pollen in sagittal section. Showing in detail the microscopic structure. On a green base with description key. In one piece.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.