ContactUsLinkComponent
Solid Alcohol and Esterification Lab Activity
Solid Alcohol and Esterification Lab Activity
Catalog # 470007-770
Supplier:  Lab-Aids
undefined
Solid Alcohol and Esterification Lab Activity
Catalog # 470007-770
Supplier:  Lab-Aids
Supplier Number:  113
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Solid Alcohol and Esterification Lab Activity
  • No. of lab groups:
    12
  • Cat. No.:
    470007-770
  • Includes:
    Teacher Guide, SDS, Student Guide/Worksheets
  • Supplier no.:
    113
Specifications
About this item
Synthesize Chemical Substances Normally Found in Nature
  • Consists of four lab activities
  • Synthesize chemical substances found in nature
  • Uses 4 to 6 50 minute class periods to complete
Create a bridge from organic chemical concepts to their practical applications. At the beginning of the lab, students make their own heating unit. Then they’ll ignite a solid alcohol to provide energy for synthesizing three different esters. Students will produce ethyl acetate, methyl salicylate (oil of winter green), and amyl acetate (banana oil). They’ll also create a molecular model of ethyl acetate using molecular model parts.
Ordering information: Kit includes set of molecular model parts, Aluminum sheets, Metal pots, Test tubes, Calibrated tubes, Wood splints, Drierite, Ethanol, Glacial Acetic Acid, Salicylic Acid, Calcium Acetate Solution, Amyl Alcohol, and Dilute Sulfuric Acid Methanol. Test tube holders are needed but not included.