Soil/Sediment Sifter
Soil/Sediment Sifter
Catalog # 470014-728
Supplier #: 621-7120
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SOIL SEDIMENT SIFTER 24X15X1 IN 2MM MESH Catalog #470356-314
Product 470356-314 is a replacement for 470014-728 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Ideal for soil and general particle sifting, this sifter’s 2mm aperture (2000 micron) corresponds to sieve size 10. The high-quality, durable wood frame and steel mesh will last through years of student use. Size: 24" x 16" x 3".
Specifications
- Description:Soil/Sediment Sifter