To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Sodium Dichromate Dihydrate
- Size:500 g
- Chemical/physical specifications:Crystal
- Cat. No.:470302-532
- Grade:Laboratory
- Supplier no.:SS0520-500G
Specifications
About this item
CAS Number: 7789-12-0
Formula Weight: 298.05
Formula: Na2Cr2O7·2H2O
Hazard Info: Toxic, Corrosive
Density (g/mL): 2.348
Solubility: Water
Synonyms: Sodium Bichromate
Shelf Life (months): 12
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.