Orders LinkContactUsLinkComponent
Snake Skull
Snake Skull, 1, 1", 1 .5", 1"
  470027-946
 :  WSS-505
Snake Skull
  470027-946
Snake Skull
Snake Skull
Snake Skull, 1, 1", 1 .5", 1"
$154.95
Each
A specimen of a common nonvenomous genus such as Thamnophis, this large, unmounted snake skull is ideal for up-close examination; its hinged jaws can be used to demonstrate how snakes swallow large prey.
  • Natural bone
  • Professionally cleaned and prepared
 : This is a real specimen, so size and appearance may vary from the image.
  • Description:
    Snake Skull
  • Height:
    1"
  • Length:
    1 .5"
  • Width:
    1"
  • No. of parts:
    1