ContactUsLinkComponent
Slide Holder Strip
Slide Holder Strip
Catalog # 470201-794
Supplier:  Bel-Art
undefined
Slide Holder Strip
Catalog # 470201-794
Supplier:  Bel-Art
Supplier Number:  F44542-0000_SE
Specifications
  • Description:
    Microscope Slide Holder Strip
  • Cat. No.:
    470201-794
  • No. of places:
    10
  • Supplier no.:
    445420000
Specifications
About this item
Rest Slides Vertically for Drying or Quick Retrieval
  • Quality slide making supplies
  • Dry slides evenly in one location
The plastic holder supports up to ten slides of varying widths and it has a front support ledge for drying slides.