Order Entry
United States
ContactUsLinkComponent
The Skin Poster
The Skin Poster
  470112-298
 :  Avantor
undefined
The Skin Poster
  470112-298
 :  Avantor

 

  • Width:
    50 cm
  • Description:
    Skin Poster
  • Length:
    66 cm
  • Cat. No.:
    470112-298
  • Finish:
    Laminated
  • Supplier no.:
    1587796139

 

 

Poster shows epidermis and dermis, and subcutaneous layer of the skin.

  • Detailed cross-section

Other drawings include hair root, fingernail, acne, fever blisters, frostbite and warts.