Specifications
- Description:Simulation of HIV Detection by Western Blot Kit
- No. of lab groups:6
- Cat. No.:470120-908
- Supplier no.:275
Specifications
About this item
This kit provides a hands-on examination of immunology procedures.
- Students Practice Immunology Procedures
- Time Required: ~75 min over Two Days
- Instructions Included
The second assay used to confirm a positive HIV ELISA result is the Western Blot.
Students separate protein samples from hypothetical patients on agarose gels, transfer the samples to a membrane, and detect the simulated HIV proteins.