Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Shigella sonnei
- Cat. No.:470179-154
- Packaging:Tube
- Media:Nutrient agar
- Supplier no.:WARD470179-154
- Incubation conditions:37 °C
Specifications
About this item
isolated from human intestine. Lactose positive and sucrose positive after a few days’ growth. Causes a mild form of dysentery.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Motility: Negative
- Morphology: Rods
- Gram Reaction: Negative
- Respiration: Facultatively Anaerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout