Self-Zeroing Automatic Buret Kit
Catalog # 470211-802
Supplier: Thermo Fisher Scientific
Specifications
- Description:Self-Zeroing Automatic Buret Kit
- Cat. No.:470211-802
- Supplier no.:3645-2550
Specifications
About this item
Nalgene® This kit makes any 25 or 50 mL buret both auto-filling and self-zeroing. With 1000 mL squeeze bottle, 1.2 m of clear flexible tubing and fill tube with adapter. Buret not included.