Orders LinkContactUsLinkComponent
Ward's® Seed Germination Kit
Ward's® Seed Germination Kit
WARD’S Seed Germination Kit
  470030-302
 :   Avantor
Ward's® Seed Germination Kit
  470030-302
 :  Avantor
Ward's® Seed Germination Kit
Ward's® Seed Germination Kit
WARD’S Seed Germination Kit
$15.99
KIT(5items) Kit of 5
Observe environmental effects on germination.
Vary environmental conditions such as moisture, temperature, and oxygen levels, and then germinate the provided seeds. Students will be able to determine the seeds’ adaptability and hardiness, as well as study the initiation of plant growth in monocots (corn), dicots (bean, pea, radish), and gymnosperms (pine). Instructions are included. Quantity: 100 seeds of each type. Set of five.
  • Description:
    WARD’S Seed Germination Kit