ContactUsLinkComponent
Sea Urchin, Pluteus Larva Slide
Sea Urchin, Pluteus Larva Slide
Catalog # 470182-376
Supplier:  Avantor
undefined
Sea Urchin, Pluteus Larva Slide
Catalog # 470182-376
Supplier:  Avantor
Specifications
  • Description:
    Sea Urchin, Pluteus Larva
  • Cat. No.:
    470182-376
  • Stain:
    Hematoxylin
  • Supplier no.:
    WARD470182-376
  • Orientation:
    Whole mount
Specifications
About this item
Sea Urchin Development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Free-swimming bilaterally symmetrical ciliated larva.