ContactUsLinkComponent
Sagittaria Grass Leaf
  470194-688
undefined
Sagittaria Grass Leaf
  470194-688
 :  867800
  • Description:
    Sagittaria Grass Leaf
  • Cat. No.:
    470194-688
  • Supplier no.:
    WARD470194-688
Sagittaria graminea. Monocot. Rooted aquarium plant with long, narrow leaves. Size: 5– 8". Package of six.
Unless otherwise noted, plants are supplied in waterproof, plastic bags that keep plants fresh and healthy. Care instructions are included. Be sure to check with your state’s Department of Agriculture for any restrictions.